Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0 zł
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Poland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Lactic acid
Lactic acid 10% in aqueous solution
Lactic acid 10% in aqueous solution
Lactic acid 10% in aqueous solution
CHMP115642634.1000
:
CHEMPUR
:
Lactic acid 10% in aqueous solution
CHMP115642634.1000
:
CHEMPUR
:
Pack type:
Glass bottle
Pk:
1 L
Seq:
5037
:
:
90.08 g/mol
:
200-018-0
:
GHS05
:
Ambient
:
50-21-5
:
598-82-3
:
H₃CCH(OH)COOH
:
MFCD00004520