Order Entry
Poland
ContactUsLinkComponent
5,6-Benzo-4,7,13,16,21,24-hexaoxa-1,10-diazabicyclo[8.8.8]hexacos-5-ene ([2.2.2]Benzocryptand, Kryptofix® 222B) 50% in toluene, Sigma-Aldrich®
5,6-Benzo-4,7,13,16,21,24-hexaoxa-1,10-diazabicyclo[8.8.8]hexacos-5-ene ([2.2.2]Benzocryptand, Kryptofix® 222B) 50% in toluene, Sigma-Aldrich®
Catalog # SIAL308579-100MG
Supplier:  Merck
CAS Number:  
5,6-Benzo-4,7,13,16,21,24-hexaoxa-1,10-diazabicyclo[8.8.8]hexacos-5-ene ([2.2.2]Benzocryptand, Kryptofix® 222B) 50% in toluene, Sigma-Aldrich®
Catalog # SIAL308579-100MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :M-Clarity™ quality level = MQ100
EINECS:250-532-4
CAS Number:31250-18-7
Formula:C₂₂H₃₆N₂O₆