2-Ethyl-3,5(or 6)-dimethylpyrazine (mixture of isomers), Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:27043-05-6
:
Formula:C₈H₁₂N₂
MDL Number:MFCD00047392
:136.194 g/mol
:M-Clarity™ quality level = MQ300
EINECS:248-182-2
Frequently Bought Together