Order Entry
Poland
ContactUsLinkComponent
4'-Carboxybenzo-15-crown-5 (4'-carboxybenzo-1,4,7,10,13-pentaoxacyclopentadecane), Sigma-Aldrich®
4'-Carboxybenzo-15-crown-5 (4'-carboxybenzo-1,4,7,10,13-pentaoxacyclopentadecane), Sigma-Aldrich®
Catalog # SIAL21874-1G
Supplier:  Merck
CAS Number:  
4'-Carboxybenzo-15-crown-5 (4'-carboxybenzo-1,4,7,10,13-pentaoxacyclopentadecane), Sigma-Aldrich®
Catalog # SIAL21874-1G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :312.3151 g/mol
CAS Number:56683-55-7
Formula:C₁₅H₂₀O₇
MDL Number:MFCD00143276