
1,3-Diethyl-2-thiobarbituric acid 99%
:5217-47-0
:
:C₈H₁₂N₂O₂S
:MFCD00006676
:200.260 g/mol
:106…109 °C
:Ambient
:226-010-7
:2811
:6.1,III
:GHS06
- Appearance:White to light yellow Powder or crystals
- Infrared spectrum:Conforms
- Melting point:105 to 109 °C
- GC:≥98.5 %
- Titration with NaOH:≥98.5 %
Frequently Bought Together