
Bis(pentamethylcyclopentadienyl)zirconium(IV) dichloride ≥99%
Catalog # 39555.04
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:2 g
- Seq:3107
Specifications
About this item
:
:432.582 g/mol
Storage Temperature:Ambient
UN:1759
ADR:8,II
:GHS05
CAS Number:54039-38-2
Formula:C₂₀H₃₀Cl₂Zr
MDL Number:MFCD00058849