Order Entry
Poland
Orders LinkContactUsLinkComponent
2,2-Dimethylcyclohexanone, Sigma-Aldrich®

2,2-Dimethylcyclohexanone, Sigma-Aldrich®

Glass bottle for solids, 1 g
Catalog # SIAL373613-1G
Supplier:   Merck
2,2-Dimethylcyclohexanone, Sigma-Aldrich®
Catalog # SIAL373613-1G
Supplier:  Merck
CAS Number:  1193-47-1
2,2-Dimethylcyclohexanone, Sigma-Aldrich®

Glass bottle for solids, 1 g

CAS Number:  1193-47-1
Price
574 zł
Log in for availability
1 * 1 g
Product Details & Documents
  • CAS Number:1193-47-1
  • Formula:C₈H₁₄O
  • MDL Number:MFCD00134382
  • Molecular Weight:126.1962 g/mol
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications