
2,2-Diphenylpropylamine hydrochloride ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:40691-66-5
:
Formula:C₁₅H₁₈ClN
MDL Number:MFCD00008133
:247.77 g/mol
Melting Pt:250…256 °C
EINECS:255-039-8
:GHS07
Storage Temperature:Temperatura otoczenia
Related Information
Frequently Bought Together