Potassium dichromate 0.25 mol/l (1.5 N) in aqueous solution volumetric solution
Supplier: CHEMPUR
Certificates
About this item
CAS Number:7778-50-9
:
Formula:K₂Cr₂O₇
MDL Number:MFCD00011367
:294.18 g/mol
Storage Temperature:Ambient
EINECS:231-906-6
UN:3287
ADR:6.1,III
:GHS06
:GHS08
Frequently Bought Together