Order Entry
Poland
ContactUsLinkComponent
 
Sodium deuteroxide 30% in D₂O (99% D) (Isotopic), Sigma-Aldrich®
Catalog # SIAL164488-20G
Supplier:  Merck
CAS Number:  
Sodium deuteroxide 30% in D₂O (99% D) (Isotopic), Sigma-Aldrich®
Catalog # SIAL164488-20G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    20 g
  • Seq:
    3290

Specifications

About this item

 :
 :41.01 g/mol
ADR:8,II
 :M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:237-825-2
UN:1824
CAS Number:14014-06-3
Formula:NaOD
MDL Number:MFCD00037669