Methyl phenylglyoxylate for synthesis, Sigma-Aldrich®
Catalog # 8.14269.0025
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 ml
- Seq:2315
Specifications
Specification Test Results
- Solubility:(20 °C) insoluble
- Melting Point:16 °C
- Molar Mass:164.16 g/mol
- Boiling Point:137 °C (19 hPa)
- Vapor Pressure:- 1 hPa (20 °C)
- Flash Point:77 °C
- Refractive Index:1.585 (20 °C, 589 nm)
- Density:1.16 g/cm3 (20 °C)
- Ignition Point:500 °C
- Synonyms:Phenylglyoxylic acid methyl ester, Benzoyl formic acid methyl ester
- Hill Formula:C9H8O3
- Chemical Formula:C6H5COCOOCH3
Specification Test Results
About this item
:
:164.1600 g/mol
:M-Clarity™ quality level = MQ200
EINECS:239-263-3
CAS Number:15206-55-0
Formula:C₉H₈O₃
MDL Number:MFCD00008443