Calcium chloride, anhydrous drying agent
POCH874890461-1KG
:
- Pack type:Glass bottle for solids
- Pk:1 kg
- Seq:4024
:
:14,01659
:1600 °C (1013 hPa)
:772 °C
:2.174 g/cm³ (20 °C)
:110.98 g/mol
:Ambient
:233-140-8
:01-2119494219-28
:GHS07
:10043-52-4
:CaCl₂
:MFCD00010903