Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0 zł
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Poland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Calcium chloride hexahydrate
Calcium chloride hexahydrate, pure
Calcium chloride hexahydrate, pure
Calcium chloride hexahydrate, pure
POCH875010429-1KG
:
POCH AVANTOR BRAND
:
Calcium chloride hexahydrate, pure
POCH875010429-1KG
:
POCH AVANTOR BRAND
:
Pack type:
Glass bottle for solids
Pk:
1 kg
Seq:
4024
:
:
219.08 g/mol
:
13,01660
:
29.54 °C
:
1.71 g/cm³ (25 °C)
:
233-140-8
:
GHS07
:
Ambient
:
7774-34-7
:
CaCl₂·6H₂O
:
MFCD00149616