
Sodium bisulphite 40% in aqueous solution, pure
Catalog # POCH808165428-1L
Supplier: POCH AVANTOR BRAND
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
Specifications
About this item
:
ADR:8,III
:104.06 g/mol
EINECS:231-548-0
UN:2693
:GHS07
CAS Number:7631-90-5
Formula:HNaO₃S
MDL Number:MFCD00167602