Paraffin, Solidification point: 58-60°C for microscopy
Catalog # POCH714459593-1KG
Supplier: POCH AVANTOR BRAND
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 kg
- Seq:4024
Specifications
About this item
:
EINECS:232-315-6
CAS Number:8002-74-2
:64742-51-4