Paraffin, Solidification point: 56-58°C for microscopy
Catalog # POCH714457592-1KG
Supplier: POCH AVANTOR BRAND
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 kg
- Seq:4024
Specifications
About this item
:
Density:0.9 g/cm³ (20 °C)
Flash Pt:>100 °C
EINECS:232-315-6
CAS Number:8002-74-2
:64742-51-4