Order Entry
Poland
ContactUsLinkComponent
Iron(III) chloride 40% in aqueous solution, specially pure
Corrosive
Irritant
Iron(III) chloride 40% in aqueous solution, specially pure
Catalog # POCH904190698-40KG
Supplier:  POCH AVANTOR BRAND
CAS Number:  
Corrosive
Irritant
Iron(III) chloride 40% in aqueous solution, specially pure
Catalog # POCH904190698-40KG
Supplier:  POCH AVANTOR BRAND
CAS Number:  

Specifications

  • Pk:
    40 kg
  • Seq:
    4367

Specifications

About this item

 :
 :162.21 g/mol
EINECS:231-729-4
UN:2582
ADR:8,III
 :GHS05
 :GHS07
CAS Number:7705-08-0
Formula:FeCl₃
MDL Number:MFCD00011005