Order Entry
Poland
ContactUsLinkComponent
Potassium thiocyanate 0.1 mol/l (0.1 N) in aqueous solution
Potassium thiocyanate 0.1 mol/l (0.1 N) in aqueous solution
Catalog # CHMP745523161.0001
Supplier:  CHEMPUR
CAS Number:  
Potassium thiocyanate 0.1 mol/l (0.1 N) in aqueous solution
Catalog # CHMP745523161.0001
Supplier:  CHEMPUR
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    0001

Specifications

About this item

 :
 :97.18 g/mol
Storage Temperature:Ambient
EINECS:206-370-1
CAS Number:333-20-0
Formula:KSCN
MDL Number:MFCD00011413