Order Entry
Poland
Orders LinkContactUsLinkComponent
 

Pyrocatechol, pure

Glass bottle for solids, 100 g
Catalog # POCH726000427-100G
Irritant
Supplier:   POCH AVANTOR BRAND
Pyrocatechol, pure
Catalog # POCH726000427-100G
Supplier:  POCH AVANTOR BRAND
CAS Number:  120-80-9
Pyrocatechol, pure

Glass bottle for solids, 100 g

CAS Number:  120-80-9
Price
298 zł
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  o-Dihydroxybenzene, Catechol, 1,2-Dihydroxybenzene
  • CAS Number:120-80-9
  • Formula:C₆H₆O₂
  • MDL Number:MFCD00002188
  • Molecular Weight:110.11 g/mol
  • Merck Index:13,08089
  • Boiling Pt:245 °C (1013 hPa)
  • Melting Pt:103…105 °C
  • Density:1.0548 g/cm³ (20 °C)
  • Flash Pt:127 °C
  • Storage Temperature:Ambient
  • EINECS:204-427-5
  • UN:2811
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.