

Acetone ≥99.8%, SupraSolv® for GC-MS, Supelco®
: Merck
:67-64-1
:
:CH₃COCH₃
:MFCD00008765
:58.08 g/mol
:13,00067
:56.2 °C (1013 hPa)
:–95.4 °C
:0.792 g/cm³ (20 °C)
:200-662-2
:1090
:3,II
:M-Clarity™ quality level = MQ100
:<–20 °C (closed cup)
:01-2119471330-49
:GHS02
:GHS07
:Ambient
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended after discussion
- Purity (GC):=99.8%
- Identity(IR):Conforms
- Evaporation Residue:= 3.0 mg/l
- Water:= 0.05%
- Color:=10 Hazen
- GC/MSD (retention range n-undecane to n-tetracontane, scanning area 30-600 amu, individual signals (n-tetradecane standard)):= 3ng/ml
- Suitability:Residue Analysis
Frequently Bought Together