Replacement needles for syringes, Neuros™
Supplier: HAMILTON BONADUZ
Certificates
About this item
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Collecting trays, b.safe
Supplier: Bohlender
VWR®, Crimp Neck Vials ND11
Supplier: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Lens tissues
Supplier: GLASWARENFABRIK KARL HECHT
VWR® Traceable®, Infrared Core Thermometer
Supplier: VWR Collection
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Syringes, Neuros™
Supplier: HAMILTON BONADUZ
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Replacement needles for syringes, Neuros™
Supplier: HAMILTON BONADUZ