(-)-Carveol (mixture of cis- and trans-isomers), Sigma-Aldrich®
Catalog # SIAL192384-10G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
- Seq:3236
Specifications
About this item
:
:152.24 g/mol
:M-Clarity™ quality level = MQ100
EINECS:202-757-4
CAS Number:99-48-9
:MFCD00062993
Formula:C₁₀H₁₆O
MDL Number:MFCD00869995