
(-)-Carveol (mixture of cis- and trans-isomers) ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:99-48-9
:MFCD00062993
Formula:C₁₀H₁₆O
MDL Number:MFCD00869995
:152.24 g/mol
Boiling Pt:227 °C
Density:0.96
Flash Pt:98 °C (209 °F)
EINECS:202-757-4
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together