Order Entry
Poland
ContactUsLinkComponent
5,5-Dimethyloxazolidine-2,4-dione (Dimethadione)
5,5-Dimethyloxazolidine-2,4-dione (Dimethadione)
Catalog # MOLEM33580189
Supplier:  Molekula
CAS Number:  
5,5-Dimethyloxazolidine-2,4-dione (Dimethadione)
Catalog # MOLEM33580189
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :129.12 g/mol
EINECS:211-781-4
CAS Number:695-53-4
Formula:C₅H₇NO₃
MDL Number:MFCD00005379