Order Entry
Poland
ContactUsLinkComponent
(R)-(+)-2,2',6,6'-Tetramethoxy-4,4'-bis(di(3,5-xylyl)phosphino)-3,3'-bipyridine, Sigma-Aldrich®
(R)-(+)-2,2',6,6'-Tetramethoxy-4,4'-bis(di(3,5-xylyl)phosphino)-3,3'-bipyridine, Sigma-Aldrich®
Catalog # SIAL676004-100MG
Supplier:  Merck
CAS Number:  
(R)-(+)-2,2',6,6'-Tetramethoxy-4,4'-bis(di(3,5-xylyl)phosphino)-3,3'-bipyridine, Sigma-Aldrich®
Catalog # SIAL676004-100MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :756.85 g/mol
 :M-Clarity™ quality level = MQ100
CAS Number:442905-33-1
Formula:C₄₆H₅₀N₂O₄P₂
MDL Number:MFCD04974235