Order Entry
Poland
Orders LinkContactUsLinkComponent
L(-)-Carnitine

L(-)-Carnitine

Glass bottle for solids, 25 g
Catalog # MOLEM59240682
Marketsource
Supplier:   Molekula
L(-)-Carnitine
Catalog # MOLEM59240682
Supplier:  Molekula
Marketsource
CAS Number:  541-15-1
L(-)-Carnitine

Glass bottle for solids, 25 g

CAS Number:  541-15-1
Price
Inquire For Price
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  Levocarnitine, L-Carnitine, Vitamin BT, (R)-3-Hydroxy-4-(trimethylammonio)butanoate, R)-(-)-3-Hydroxy-4-(trimethylammonio)butyrate
  • CAS Number:541-15-1
  • Formula:C₇H₁₅NO₃
  • MDL Number:MFCD00038747
  • Molecular Weight:161.20 g/mol
  • EINECS:208-768-0
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.