Order Entry
Poland
ContactUsLinkComponent
DL-Carnitine hydrochloride, Sigma-Aldrich®
DL-Carnitine hydrochloride, Sigma-Aldrich®
Catalog # SIALC9500-100G
Supplier:  Merck
CAS Number:  
DL-Carnitine hydrochloride, Sigma-Aldrich®
Catalog # SIALC9500-100G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :197.66 g/mol
 :M-Clarity™ quality level = MQ200
EINECS:207-309-1
CAS Number:461-05-2
Formula:C₇H₁₆ClNO₃
MDL Number:MFCD00011904