2-Nitrocinnamaldehyde for synthesis, Sigma-Aldrich®
Catalog # 8.18597.0010
Supplier: Merck
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:10 g
- Seq:3236
Specifications
Specification Test Results
- Solubility:(20 °C) slightly soluble
- Melting Point:126 - 129 °C
- Molar Mass:177.15 g/mol
- Hill Formula:C9H7NO3
- Chemical Formula:2-(NO2)C6H4CH=CHCHO
Specification Test Results
About this item
:
:M-Clarity™ quality level = MQ200
:177.16 g/mol
EINECS:215-988-0
CAS Number:1466-88-2
Formula:C₉H₇NO₃
MDL Number:MFCD00007188