
Sodium tetrakis[3,5-bis(trifluoromethyl)phenyl]borate, (H₂O 1-5%) ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:79060-88-1
:
Formula:C₃₂H₁₂BF₂₄Na
MDL Number:MFCD00043323
:886.20 g/mol
Melting Pt:310 °C (dec.)
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together