Order Entry
Poland
ContactUsLinkComponent
1,1-Diphenylhydrazine hydrochloride, Sigma-Aldrich®
1,1-Diphenylhydrazine hydrochloride, Sigma-Aldrich®
Catalog # SIAL114596-25G
Supplier:  Merck
CAS Number:  
1,1-Diphenylhydrazine hydrochloride, Sigma-Aldrich®
Catalog # SIAL114596-25G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :220.70 g/mol
EINECS:208-481-0
CAS Number:530-47-2
Formula:C₁₂H₁₃ClN₂
MDL Number:MFCD00012950