5-Fluoro-5'-deoxyuridine (Doxifluridine)
Catalog # MOLE10130089-25MG
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 mg
- Seq:2315
Specifications
About this item
:
:246.19 g/mol
EINECS:221-440-1
CAS Number:3094-09-5
:38817-29-7
Formula:C₉H₁₁FN₂O₅
MDL Number:MFCD00866530