Order Entry
Poland
ContactUsLinkComponent
Ethosuximide
Ethosuximide
Catalog # PROOICRS0307
CAS Number:  
Ethosuximide
Catalog # PROOICRS0307
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :141.17 g/mol
EINECS:201-048-7
CAS Number:77-67-8
Formula:C₇H₁₁NO₂
MDL Number:MFCD00072123