


Zinc sulphate 0.01 mol concentrated aqueous solution volumetric solution
Catalog # CHMP162657501.0001
Supplier: CHEMPUR
CAS Number:
Specifications
- Pk:1
Specifications
About this item
:
:161.45 g/mol
Storage Temperature:Ambient
EINECS:231-793-3
UN:3082
ADR:9,III
:GHS05
:GHS07
:GHS09
CAS Number:7733-02-0
Formula:O₄SZn
MDL Number:MFCD00011302