EDTA disodium salt 0.015 mol/l (0.03 N) in aqueous solution volumetric solution
Supplier: CHEMPUR
Certificates
About this item
CAS Number:139-33-3
:
Formula:C₁₀H₁₄N₂Na₂O₈
MDL Number:MFCD00070672
:336.21 g/mol
Storage Temperature:Ambient
EINECS:205-358-3
Frequently Bought Together