

2-Propanol, specially pure for the electronics industry
POCH751500612-1L
:
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and after discussion
:
:60.1 g/mol
:13,05228
:82 °C (1013 hPa)
:–89 °C
:0.786 g/cm³ (20 °C)
:12 °C
:Ambient
:200-661-7
:1219
:3,II
:01-2119457558-25
:GHS02
:GHS07
:67-63-0
:(CH₃)₂CHOH
:MFCD00011674