Order Entry
Poland
Orders LinkContactUsLinkComponent
 

1,2-Dichloroethane, pure analytical reagent

Glass bottle, 1 L
Catalog # POCH388862111-1L
Flammable
Health
Irritant
Supplier:   POCH AVANTOR BRAND
1,2-Dichloroethane, pure analytical reagent
Catalog # POCH388862111-1L
Supplier:  POCH AVANTOR BRAND
CAS Number:  107-06-2
1,2-Dichloroethane, pure analytical reagent
1,2-Dichloroethane, pure analytical reagent

Glass bottle, 1 L

CAS Number:  107-06-2
Price
555 zł
Log in for availability
1 * 1 L
Product Details & Documents
Synonyms:  Ethylene chloride, Ethylene dichloride, Dichloroethane
  • CAS Number:107-06-2
  • Formula:ClH₂CCH₂Cl
  • MDL Number:MFCD00000963
  • Molecular Weight:98.96 g/mol
  • Merck Index:13,03831
  • Boiling Pt:83.5 °C (1013 hPa)
  • Melting Pt:–35.5 °C
  • Density:1.254 g/cm³ (20 °C)
  • Flash Pt:13 °C
  • Storage Temperature:Ambient
  • EINECS:203-458-1
  • UN:1184
  • ADR:3,II
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L

Specifications