Butyl acetate, pure analytical reagent
POCH211500119-1L
:
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and after discussion
:
:116.16 g/mol
:13,01534
:126 °C (1013 hPa)
:–76 °C
:0.875 g/cm³ (20 °C)
:25 °C
:Ambient
:204-658-1
:1123
:3,III
:GHS02
:GHS07
:123-86-4
:CH₃COO(CH₂)₃CH₃
:MFCD00009445