0 zł
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Poland
ContactUsLinkComponent
Sulphuric acid
Sulphuric acid 0.5 mol/l (1 N) in aqueous solution
Sulphuric acid 0.5 mol/l (1 N) in aqueous solution
POCH575065166-5L
:
POCH AVANTOR BRAND
:
Sulphuric acid 0.5 mol/l (1 N) in aqueous solution
POCH575065166-5L
:
POCH AVANTOR BRAND
:
Pk:
5 L
Seq:
5190
:
:
98.08 g/mol
:
1.01 g/cm³ (20 °C)
:
Ambient
:
231-639-5
:
2796
:
8,II
:
7664-93-9
:
H₂SO₄
:
MFCD00064589