


Formaldehyde 36 - 38% in aqueous solution, pure analytical reagent
Supplier: CHEMPUR
Certificates
About this item
CAS Number:50-00-0
:
Formula:CH₂O
MDL Number:MFCD00003274
:30.03 g/mol
Storage Temperature:Ambient
EINECS:200-001-8
UN:2209
ADR:8,III
:GHS06
:GHS05
:GHS08
Frequently Bought Together