Order Entry
Poland
ContactUsLinkComponent
Formaldehyde 20% in aqueous solution, specially pure
Toxic
Health
Formaldehyde 20% in aqueous solution, specially pure
Catalog # POCH432170694-1L
Supplier:  POCH AVANTOR BRAND
CAS Number:  
Toxic
Health
Formaldehyde 20% in aqueous solution, specially pure
Catalog # POCH432170694-1L
Supplier:  POCH AVANTOR BRAND
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :30.03 g/mol
Storage Temperature:Ambient
EINECS:200-001-8
UN:3334
ADR:9,
 :GHS06
 :GHS08
CAS Number:50-00-0
Formula:CH₂O
MDL Number:MFCD00003274