

Xylene (mixture of isomers), pure analytical reagent
Catalog # CHMP115086037.1000
Supplier: CHEMPUR
CAS Number:
Specifications
- Pk:1 L
Specifications
About this item
:
:106.17 g/mol
Merck Index:13,10136
Boiling Pt:140 °C (1013 hPa)
Melting Pt:–34 °C
Density:0.86…0.88 g/cm³ (20 °C)
Flash Pt:25 °C
Storage Temperature:Ambient
EINECS:215-535-7
UN:1307
ADR:3,III
:GHS02
:GHS07
CAS Number:1330-20-7
Formula:C₆H₄(CH₃)₂
MDL Number:MFCD00077264