Heptane (mixture of isomers), pure analytical reagent
POCH470470117-1L
:
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
:
:13,04679
:97…98 °C (1013 hPa)
:–90.5 °C
:0.68 g/cm³ (20 °C)
:–4 °C
:Ambient
:205-563-8
:1206
:3,II
:100.2 g/mol
:GHS02
:GHS08
:GHS07
:GHS09
:142-82-5
:H₃C(CH₂)₅CH₃
:MFCD00009544