
Dithizone indicator
Supplier: CHEMPUR
Certificates
About this item
CAS Number:60-10-6
:
Formula:C₆H₅NHNHCSN=NC₆H₅
MDL Number:MFCD00003025
:256.33 g/mol
Merck Index:13,03413
Boiling Pt:406 °C (1013 hPa)
Melting Pt:168 °C
EINECS:200-454-1
:GHS07
Frequently Bought Together