Order Entry
Poland
ContactUsLinkComponent
Molecular sieve A4 (0.4 nm, 4 Å)
Molecular sieve A4 (0.4 nm, 4 Å)
Catalog # POCH787500468-500G
Supplier:  POCH AVANTOR BRAND
CAS Number:  
Molecular sieve A4 (0.4 nm, 4 Å)
Catalog # POCH787500468-500G
Supplier:  POCH AVANTOR BRAND
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g
  • Seq:
    3645

Specifications

About this item

 :
Density:1.1…2.2 g/cm³ (20 °C)
CAS Number:70955-01-0
 :1318-02-1
MDL Number:MFCD03457537