Chloroform stabilised, specially pure for photography
Catalog # POCH234434429-1L
Supplier: POCH AVANTOR BRAND
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
- Environmentally Preferable:
Specifications
About this item
Synonyms: Methane, trichloro-, Trichloroform, Methylidyne trichloride, Trichloromethane
:
:119.38 g/mol
EINECS:200-663-8
UN:1888
REACH:05-2114614906-45
:GHS06
:GHS08
Storage Temperature:Ambient
CAS Number:67-66-3
Formula:CHCl₃
MDL Number:MFCD00000826
Boiling Pt:61.7
Melting Pt:–63 °C
Density:1.4832 g/cm³ (20 °C)
ADR:6.1 III
Merck Index:13,02160