

Copper(II) chloride, anhydrous, pure
Catalog # POCH657280428-250G
Supplier: POCH AVANTOR BRAND
CAS Number:
Specifications
- Pk:250 g
- Seq:3545
Specifications
About this item
:
:134.45 g/mol
Merck Index:13,02660
Boiling Pt:1365.85 °C (1013 hPa)
Melting Pt:628 °C
Density:3.054 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-210-2
UN:2802
ADR:8,III
:GHS07
:GHS09
CAS Number:7447-39-4
Formula:Cl₂Cu
MDL Number:MFCD00010972