D-(+)-Glucose, anhydrous, pure
Supplier: POCH AVANTOR BRAND
Certificates
About this item
CAS Number:50-99-7
:492-62-6
Formula:C₆H₁₂O₆
MDL Number:MFCD00148912
Merck Index:14,04459
Melting Pt:146 °C
Density:1.54 g/cm³ (20 °C)
:180.16 g/mol
Storage Temperature:Ambient
EINECS:200-075-1
Frequently Bought Together