Ammonium thiocyanate, pure analytical reagent
:1762-95-4
:
:NH₄SCN
:MFCD00011428
:76.12 g/mol
:13,00564
:150 °C
:1.305 g/cm³ (20 °C)
:217-175-6
:GHS07
:Ambient
Frequently Bought Together
Ammonium acetate ≥97%, GPR RECTAPUR®
: VWR Chemicals
Calcium chloride dihydrate 99-105% ACS
: VWR Chemicals
Potassium dihydrogen phosphate ≥99%, powder, GPR RECTAPUR®
: VWR Chemicals
Ammonium thiocyanate 99+%, extra pure
: Thermo Fisher Scientific
POTASU JODEK CZDA OP. 500 G 1 * 500 g
: POCH AVANTOR BRAND
Iron, powder, Sigma-Aldrich®
: Merck
KWAS SOLNY 35 - 38% CZ OP. 5 DM3 1 * 5 L
: POCH AVANTOR BRAND