Order Entry
Poland
ContactUsLinkComponent
3,3',5,5'-Tetramethylbenzidine dihydrochloride hydrate, Sigma-Aldrich®
3,3',5,5'-Tetramethylbenzidine dihydrochloride hydrate, Sigma-Aldrich®
Catalog # SIAL861510-1G
Supplier:  Merck
CAS Number:  
3,3',5,5'-Tetramethylbenzidine dihydrochloride hydrate, Sigma-Aldrich®
Catalog # SIAL861510-1G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :313.27 g/mol (anhy)
 :M-Clarity™ quality level = MQ200
EINECS:264-769-6
CAS Number:207738-08-7
Formula:C₁₆H₂₀N₂·2HCl·xH₂O
MDL Number:MFCD00150104