Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0 zł
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Poland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
n-Tetradecyl-β-D-maltoside
n-Tetradecyl-β-D-maltoside, Sigma-Aldrich®
n-Tetradecyl-β-D-maltoside, Sigma-Aldrich®
n-Tetradecyl-β-D-maltoside, Sigma-Aldrich®
Catalog # SIAL15826-1G
Supplier:
Merck
CAS Number:
Expand image
n-Tetradecyl-β-D-maltoside, Sigma-Aldrich®
Catalog # SIAL15826-1G
Supplier:
Merck
CAS Number:
Specifications
Pack type:
Glass bottle for solids
Pk:
1 g
Seq:
3047
Specifications
About this item
Product Details
:
CAS Number:
18449-82-6
Formula:
C₂₆H₅₀O₁₁
Product Details